Anti-POFUT1 Mouse Monoclonal Antibody [clone: 3H1]
Lieferant: Abnova
ABNOH00023509-M01EA 473 CHF
ABNOH00023509-M01
Anti-POFUT1 Mouse Monoclonal Antibody [clone: 3H1]
Antikörper
Type: Primary
Antigen: POFUT1
Clonality: Monoclonal
Clone: 3H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human