Anti-DAAM2 Rabbit Polyclonal Antibody
Lieferant: Abnova
ABNOH00023500-D01PEA 456 CHF
ABNOH00023500-D01P
Anti-DAAM2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DAAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human