Anti-MACF1 Mouse Monoclonal Antibody [clone: 6G7]

Lieferant: Abnova
ABNOH00023499-M01EA 473 CHF
ABNOH00023499-M01
Anti-MACF1 Mouse Monoclonal Antibody [clone: 6G7]
Antikörper
Type: Primary
Antigen: MACF1
Clonality: Monoclonal
Clone: 6G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR