Anti-RIPPLY2 Mouse Monoclonal Antibody [clone: OTI1B5]

Supplier: OriGene
ORIGTA501512EA 723 CHF
ORIGTA501512
Anti-RIPPLY2 Mouse Monoclonal Antibody [clone: OTI1B5]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: RIPPLY2 (ripply transcriptional repressor 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR