Anti-MIOX Mouse Monoclonal Antibody [clone: OTI1D1]

Supplier: OriGene
ORIGTA501476EA 676 CHF
ORIGTA501476
Anti-MIOX Mouse Monoclonal Antibody [clone: OTI1D1]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: MIOX (myo-inositol oxygenase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR