Anti-EFNA2 Mouse Monoclonal Antibody [clone: OTI4C3]

Supplier: OriGene
ORIGTA501457EA 702 CHF
ORIGTA501457
Anti-EFNA2 Mouse Monoclonal Antibody [clone: OTI4C3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: EFNA2 (ephrin-A2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR