Anti-L1CAM Mouse Monoclonal Antibody [clone: OTI1D12]

Supplier: OriGene
ORIGTA501440EA 702 CHF
ORIGTA501440
Anti-L1CAM Mouse Monoclonal Antibody [clone: OTI1D12]
Antibodies
  • Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: L1CAM (L1 cell adhesion molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR