Anti-IL36A Mouse Monoclonal Antibody [clone: OTI1A4]

Supplier: OriGene
ORIGTA501407EA 702 CHF
ORIGTA501407
Anti-IL36A Mouse Monoclonal Antibody [clone: OTI1A4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: IL36A (interleukin 36, alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR