Anti-DHPS Mouse Monoclonal Antibody [clone: OTI2C9]

Supplier: OriGene
ORIGTA501393EA 708 CHF
ORIGTA501393
Anti-DHPS Mouse Monoclonal Antibody [clone: OTI2C9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: DHPS (deoxyhypusine synthase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR