Anti-HIBCH Mouse Monoclonal Antibody [clone: OTI3D11]

Supplier: OriGene
ORIGTA501387EA 676 CHF
ORIGTA501387
Anti-HIBCH Mouse Monoclonal Antibody [clone: OTI3D11]
Antibodies
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HIBCH (3-hydroxyisobutyryl-CoA hydrolase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR