Anti-C9orf41 Mouse Monoclonal Antibody [clone: OTI4B7]

Supplier: OriGene
ORIGTA501308EA 702 CHF
ORIGTA501308
Anti-C9orf41 Mouse Monoclonal Antibody [clone: OTI4B7]
Antibodies
  • Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C9orf41 (chromosome 9 open reading frame 41)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR