Anti-KHK Mouse Monoclonal Antibody [clone: OTI3C4]

Supplier: OriGene
ORIGTA501247EA 717 CHF
ORIGTA501247
Anti-KHK Mouse Monoclonal Antibody [clone: OTI3C4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: KHK (ketohexokinase (fructokinase))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR