Anti-GPI Mouse Monoclonal Antibody [clone: OTI8G7]

Supplier: OriGene
ORIGTA501135EA 717 CHF
ORIGTA501135
Anti-GPI Mouse Monoclonal Antibody [clone: OTI8G7]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: GPI (glucose-6-phosphate isomerase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR