Anti-PIM2 Mouse Monoclonal Antibody [clone: OTI8H10]

Supplier: OriGene
ORIGTA501070EA 676 CHF
ORIGTA501070
Anti-PIM2 Mouse Monoclonal Antibody [clone: OTI8H10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: PIM2 (pim-2 oncogene)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR