Anti-NPR3 Mouse Monoclonal Antibody [clone: OTI4A1]

Supplier: OriGene
ORIGTA501046EA 702 CHF
ORIGTA501046
Anti-NPR3 Mouse Monoclonal Antibody [clone: OTI4A1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: NPR3 (natriuretic peptide receptor 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR