Anti-LIPG Mouse Monoclonal Antibody [clone: OTI4C10]

Supplier: OriGene
ORIGTA501032EA 702 CHF
ORIGTA501032
Anti-LIPG Mouse Monoclonal Antibody [clone: OTI4C10]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: LIPG (lipase, endothelial)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR