Anti-PTPRE Mouse Monoclonal Antibody [clone: OTI3H8]

Supplier: OriGene
ORIGTA501029EA 717 CHF
ORIGTA501029
Anti-PTPRE Mouse Monoclonal Antibody [clone: OTI3H8]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: PTPRE (protein tyrosine phosphatase, receptor type, E)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR