Anti-MTRF1L Mouse Monoclonal Antibody [clone: OTI1B3]

Supplier: OriGene
ORIGTA501013EA 723 CHF
ORIGTA501013
Anti-MTRF1L Mouse Monoclonal Antibody [clone: OTI1B3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: MTRF1L (mitochondrial translational release factor 1-like)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR