Anti-APP Mouse Monoclonal Antibody [clone: OTI2B10]

Supplier: OriGene
ORIGTA500973EA 708 CHF
ORIGTA500973
Anti-APP Mouse Monoclonal Antibody [clone: OTI2B10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:50

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR