Anti-CELF1 Mouse Monoclonal Antibody [clone: OTI5B8]

Supplier: OriGene
ORIGTA500951EA 723 CHF
ORIGTA500951
Anti-CELF1 Mouse Monoclonal Antibody [clone: OTI5B8]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: CELF1 (CUGBP, Elav-like family member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR