Anti-SRR Mouse Monoclonal Antibody [clone: OTI1D7]

Supplier: OriGene
ORIGTA500942EA 717 CHF
ORIGTA500942
Anti-SRR Mouse Monoclonal Antibody [clone: OTI1D7]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100

Type: Primary
Antigen: SRR (serine racemase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR