Anti-CD36 Mouse Monoclonal Antibody [clone: OTI4H7]

Supplier: OriGene
ORIGTA500935EA 717 CHF
ORIGTA500935
Anti-CD36 Mouse Monoclonal Antibody [clone: OTI4H7]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR