Anti-HDHD2 Mouse Monoclonal Antibody [clone: OTI2C12]

Supplier: OriGene
ORIGTA500914EA 717 CHF
ORIGTA500914
Anti-HDHD2 Mouse Monoclonal Antibody [clone: OTI2C12]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: HDHD2 (haloacid dehalogenase-like hydrolase domain containing 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR