Anti-GBE1 Mouse Monoclonal Antibody [clone: OTI4A1]

Supplier: OriGene
ORIGTA500846EA 717 CHF
ORIGTA500846
Anti-GBE1 Mouse Monoclonal Antibody [clone: OTI4A1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: GBE1 (glucan (1,4-alpha-), branching enzyme 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR