Anti-PSMD10 Mouse Monoclonal Antibody [clone: OTI3F6]

Supplier: OriGene
ORIGTA500781EA 708 CHF
ORIGTA500781
Anti-PSMD10 Mouse Monoclonal Antibody [clone: OTI3F6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100

Type: Primary
Antigen: PSMD10 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 10)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR