Anti-RNF144B Mouse Monoclonal Antibody [clone: OTI6B5]

Supplier: OriGene
ORIGTA500705EA 717 CHF
ORIGTA500705
Anti-RNF144B Mouse Monoclonal Antibody [clone: OTI6B5]
Antibodies
Recommended Dilutions: Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: RNF144B (ring finger protein 144B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR