Anti-LTA4H Mouse Monoclonal Antibody [clone: OTI5C10]

Supplier: OriGene
ORIGTA500637EA 676 CHF
ORIGTA500637
Anti-LTA4H Mouse Monoclonal Antibody [clone: OTI5C10]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2-4μg/mg

Type: Primary
Antigen: LTA4H (leukotriene A4 hydrolase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR