Anti-ZFP36 Mouse Monoclonal Antibody [clone: OTI3D10]

Supplier: OriGene
ORIGTA500621EA 702 CHF
ORIGTA500621
Anti-ZFP36 Mouse Monoclonal Antibody [clone: OTI3D10]
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: ZFP36 (ZFP36 ring finger protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR