Anti-PPP5C Mouse Monoclonal Antibody [clone: OTI3G1]

Supplier: OriGene
ORIGTA500597EA 717 CHF
ORIGTA500597
Anti-PPP5C Mouse Monoclonal Antibody [clone: OTI3G1]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: PPP5C (protein phosphatase 5, catalytic subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR