Anti-SLC2A5 Mouse Monoclonal Antibody [clone: OTI11E1]

Supplier: OriGene
ORIGTA500574EA 702 CHF
ORIGTA500574
Anti-SLC2A5 Mouse Monoclonal Antibody [clone: OTI11E1]
Antibodies
Recommended Dilutions: Western Blot: 1:5000; Immunofluorescence: 1:100

Type: Primary
Antigen: SLC2A5 (solute carrier family 2 (facilitated glucose/fructose transporter), member 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR