Anti-SERPINA1 Mouse Monoclonal Antibody [clone: OTI9A1]

Supplier: OriGene
ORIGTA500374EA 717 CHF
ORIGTA500374
Anti-SERPINA1 Mouse Monoclonal Antibody [clone: OTI9A1]
Antibodies
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: SERPINA1 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR