Anti-CYP7B1 Mouse Monoclonal Antibody [clone: OTI7H10]

Supplier: OriGene
ORIGTA500244EA 702 CHF
ORIGTA500244
Anti-CYP7B1 Mouse Monoclonal Antibody [clone: OTI7H10]
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CYP7B1 (cytochrome P450, family 7, subfamily B, polypeptide 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR