Anti-PTCH1 Mouse Monoclonal Antibody [clone: OTI3c2]

Supplier: OriGene
ORIGTA500122EA 702 CHF
ORIGTA500122
Anti-PTCH1 Mouse Monoclonal Antibody [clone: OTI3c2]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: PTCH1 (patched 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR