Anti-SERPINA1 Mouse Monoclonal Antibody [clone: OTI1C2]

Supplier: OriGene
ORIGTA500068EA 717 CHF
ORIGTA500068
Anti-SERPINA1 Mouse Monoclonal Antibody [clone: OTI1C2]
Antibodies
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: SERPINA1 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR