Anti-EPCAM Mouse Monoclonal Antibody [clone: OTI4G10]

Supplier: OriGene
ORIGTA500055EA 702 CHF
ORIGTA500055
Anti-EPCAM Mouse Monoclonal Antibody [clone: OTI4G10]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR