Anti-SNX9 Mouse Antibody

Supplier: OriGene
ORIGTA400029EA 741 CHF
ORIGTA400029
Anti-SNX9 Mouse Antibody
Antibodies
Type: Primary
Antigen: SNX9 (sorting nexin 9)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR