Anti-BIRC5 Mouse Antibody

Supplier: OriGene
ORIGTA400024EA 741 CHF
ORIGTA400024
Anti-BIRC5 Mouse Antibody
Antibodies
Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR