Anti-CD36 Mouse Antibody

Supplier: OriGene
ORIGTA400016EA 726 CHF
ORIGTA400016
Anti-CD36 Mouse Antibody
Antibodies
Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR