Anti-CTSK Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA318059EA 1200 CHF
ORIGTA318059
Anti-CTSK Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 2.5 µg/ml; Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CTSK (cathepsin K)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR