Anti-GOT1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA318035EA 775 CHF
ORIGTA318035
Anti-GOT1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:4000; Immunochemistry: 2.5-5 µg/ml; Western Blot: 0.01-0.1 µg/ml

Type: Primary
Antigen: GOT1 (glutamic-oxaloacetic transaminase 1, soluble)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR