Anti-DRD1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA317650EA 775 CHF
ORIGTA317650
Anti-DRD1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 10 µg/ml

Type: Primary
Antigen: DRD1 (dopamine receptor D1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR