Anti-USP2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA317489EA 775 CHF
ORIGTA317489
Anti-USP2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 5 µg/ml

Type: Primary
Antigen: USP2 (ubiquitin specific peptidase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR