Anti-UBA6 Mouse Monoclonal Antibody [clone: 1D11]

Supplier: OriGene
ORIGTA317480EA 1010 CHF
ORIGTA317480
Anti-UBA6 Mouse Monoclonal Antibody [clone: 1D11]
Antibodies
Recommended Dilutions: Immunochemistry: 5 µg/ml;

Type: Primary
Antigen: UBA6 (ubiquitin-like modifier activating enzyme 6)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR