Anti-SLC11A2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA317306EA 775 CHF
ORIGTA317306
Anti-SLC11A2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 15-20 µg/ml

Type: Primary
Antigen: SLC11A2 (solute carrier family 11 (proton-coupled divalent metal ion transporter), member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR