Anti-RORB Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA317258EA 791 CHF
ORIGTA317258
Anti-RORB Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 20 µg/ml

Type: Primary
Antigen: RORB (RAR-related orphan receptor B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR