Anti-PTGDR2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA317197EA 775 CHF
ORIGTA317197
Anti-PTGDR2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 3-6 µg/ml

Type: Primary
Antigen: PTGDR2 (prostaglandin D2 receptor 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR