Anti-LPAR4 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA316967EA 791 CHF
ORIGTA316967
Anti-LPAR4 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 5-10 µg/ml

Type: Primary
Antigen: LPAR4 (lysophosphatidic acid receptor 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR