Anti-GPR183 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA316825EA 775 CHF
ORIGTA316825
Anti-GPR183 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 10 µg/ml

Type: Primary
Antigen: GPR183 (G protein-coupled receptor 183)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR