Anti-FPR2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA316750EA 775 CHF
ORIGTA316750
Anti-FPR2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 4-11 µg/ml

Type: Primary
Antigen: FPR2 (formyl peptide receptor 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR