Anti-ATF2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA316504EA 656 CHF
ORIGTA316504
Anti-ATF2 Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:20000; Immunoprecipitation: Various Dilution

Type: Primary
Antigen: ATF2 (activating transcription factor 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR